Tisk…
Electronic Battery Crimping Tools
Dodavatel: Trajan Scientific and Medical
6A08C0 6A11C0 6A11D0 6A13C0 6A13D0 6A13F0 6A20C0 6A20D0 6A20F0
SGEA6A08C0EA 30300 CZK
SGEA6A08C0 SGEA6A11C0 SGEA6A11D0 SGEA6A13C0 SGEA6A13D0 SGEA6A13F0 SGEA6A20C0 SGEA6A20D0 SGEA6A20F0
Electronic Battery Crimping Tools
Kleště krimpovací a odkrimpovací
Electronic battery crimping tools effortlessly crimp/decap aluminum and 2-part aluminum/steel caps with just the push of a button.
  • Consistently crimp caps to a reproducible seal setting with less physical strain
  • Powerful brushless motor with a fast cycle
  • Long lasting lithium ion battery provides up to 800 crimps/decaps per charge with an average recharge time of 1 to 2 hours
  • Innovative charging system, which includes a warning light when recharging is necessary
  • Can be operated while charging
  • The controls to increase or decrease crimp settlings are
More Product Information