Tisk…
Graphite GC Ferrules
Dodavatel: Trajan Scientific and Medical
211100 211103 211104 211105 211108 211110 211112 211125 211160 211164 211165 211168 211170 211174 211175 211178 2111XX 211200 211201 211204 211205 211208 211210 2112XX 211400 211405 211406 211408 211410 211420 211440 2114XX 211500 211550 211982-10 211983-10 211984-10 211988-10 211120 211600 211800 211904 211905 211908 211164-200 211406NL
SGEA211100EA 1160 CZK
SGEA211100 SGEA211103 SGEA211104 SGEA211105 SGEA211108 SGEA211110 SGEA211112 SGEA211125 SGEA211160 SGEA211164 SGEA211165 SGEA211168 SGEA211170 SGEA211174 SGEA211175 SGEA211178 SGEA2111XX SGEA211200 SGEA211201 SGEA211204 SGEA211205 SGEA211208 SGEA211210 SGEA2112XX SGEA211400 SGEA211405 SGEA211406 SGEA211408 SGEA211410 SGEA211420 SGEA211440 SGEA2114XX SGEA211500 SGEA211550 SGEA211982-10 SGEA211983-10 SGEA211984-10 SGEA211988-10 SGEA211120 SGEA211600 SGEA211800 SGEA211904 SGEA211905 SGEA211908 SGEA211164-200 SGEA211406NL
Graphite GC Ferrules
Fitinky a konektory
Graphite ferrules are commonly used in GC systems on the inlet and detector ends because they don’t stick to the column, and can be removed and reinserted easily. They should not be used with GC/MS instruments because of permeability to air.
  • Made from high purity exfoliated graphite
  • Seals with minimal torque and can be frequently reused if not over compressed
  • Graphite ferrules do not shrink in use but are very soft and must be packed and handled with care
You should always trim a short piece of the column after replacing the ferrule to be sure thatMore Product Information