Tisk…
Gas purifiers
Dodavatel: Trajan Scientific and Medical
1035210 1035220 1035230 1035250 1035158 1035154 1035168 1035164
SGEA1035210EA 5350 CZK
SGEA1035210 SGEA1035220 SGEA1035230 SGEA1035250 SGEA1035158 SGEA1035154 SGEA1035168 SGEA1035164
Gas purifiers
Filtry Filtry na plyny
Gas purifiers are designed to provide fast stabilisation times to reduce gas consumption and provide clean gas to GC and GC/MS systems.
  • Removal of oxygen, moisture and hydrocarbons that can impact analytical results
  • Fast stabilisation times
  • Improved trace analysis
  • Full removal of gas impurities for different applications
Gas purifiers are an essential part of your GC analysis as contaminants in gases can significantly impact the quality of results. Oxygen, hydrocarbons and moisture can lead to problems such as noisy baselines, moisture entering the GCMore Product Information
Objednat
Produktová specifikace
Environmentally Preferable
Velikost fitinky 1/8" 1/4''