Tisk…
Low temperature Pt100 probe
Dodavatel: SenseAnywhere
0096647494821
SENA0096647494821EA 4110 CZK
SENA0096647494821
Low temperature Pt100 probe
Čidla teplotní
This smart probe featuring a digital SenseAnywhere Bus interface has been especially designed to be used in ultra-low environments for monitoring cryogenic applications down to -200 °C. The class A wire wound ceramic Pt100 sensor ensures accurate measurements over a wide temperature range. Due to the digital interface this probe with its own serial number and integrated electonics can be calibrated. Calibrated SAB sensors can be easily hot swapped offering a big advantage and cost savings for recalibration of sensors.
  • Accurate temperature measurement
  • Realtime: 24/7 data gives a
More Product Information