Tisk…
Securibox Containers, Round, PP
Dodavatel: SEMADENI
7699
SEMA7699EA 0 CZK
SEMA7699
Securibox Containers, Round, PP
Krabice Boxy skladovací
Universal containers with snap-on closure system and tamper-evident seal.
Comprehensive container system from 17 ml to 1,8 L, comprising a white container (PP) and white snap-on closure (PE-LD) with warranty ring. The ring is easy to open. The containers can be filled quickly and easily, and also allow for quick and easy fully automated filling. Securibox can easily be resealed at any time.