Tisk…
Closures for bottles, with seal ring, PP
Dodavatel: SEMADENI
466191 471003
SEMA466191EA 10180 CZK
SEMA466191 SEMA471003
Closures for bottles, with seal ring, PP
Uzávěry Uzávěry k láhvím
PP closure, with PE sealing ring
  • Safety closures for Inter pharmaceutical glasses
  • Low water vapor permeability