Tisk…
Bottles, narrow neck, round or square, HDPE, with screw caps
Dodavatel: SEMADENI
4585 4291
SEMA4585EA 2610 CZK
SEMA4585 SEMA4291
Bottles, narrow neck, round or square, HDPE, with screw caps
Láhve Láhve plastové
HDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance
  • Un approved 3H1/X250 for fluids
With tamper evident screw cap.