Tisk…
Magnetic stirring bars, cylindrical
Dodavatel: SEMADENI
2209 0244 0257 0258 0259 0320
SEMA2209EA 693 CZK
SEMA2209 SEMA0244 SEMA0257 SEMA0258 SEMA0259 SEMA0320
Magnetic stirring bars, cylindrical
Míchadla Míchadla magnetická
Magnetic stirrer bars with permanent magnetic core and white PTFE coating.
  • Chemical resistance
  • Smooth surface and round ends
  • Temperature resistance upto 275 °C