Tisk…
Culture tubes with screw cap
Dodavatel: schuett-biotec GmbH
3561253 3563423 3560103 3563623 3562123 3562143 3563143 3563523 3561103
391-0156EA 10420 CZK
391-0156 391-0157 391-6135 SCUT3563623 SCUT3562123 SCUT3562143 SCUT3563143 SCUT3563523 SCUT3561103
Culture tubes with screw cap
Zkumavky Zkumavky kultivační
Tubes made of borosilicate with aluminium or plastic screw caps and rubber seal.
  • Heavy walled
  • Easy to inoculate and clean