Tisk…
HPLC Supply and Waste Set, V2.0
Dodavatel: S.C.A.T.
307337 307447
SCTV307337EA 17060 CZK
SCTV307337 SCTV307447
HPLC Supply and Waste Set, V2.0
Chromatografické potřeby Chromatography Spare Parts
The HPLC Supply and Waste Disposal Set V2.0 is the ultimate and universal set of all required components for solvent supply and waste of all HPLC/UHPLC systems.
  • Safety for solvent supply and waste
  • Fits every HPLC/UHPLC system
  • Including solvent bottles and waste container
  • Blocks solvent vapours
  • Protects your system from contamination
  • Keeps solvent mixtures stable
The safety set reliably connects hoses and blocks hazardous vapors. The set includes all components required for the supply (SafetyCaps, air valves and laboratory bottles) as well as theMore Product Information