Tisk…
HPLC waste-sets, V2.0
Dodavatel: S.C.A.T.
307307 307310 307328 307367 307368 307369 307370 307371 307372 307373 307324 307355 307366 307374 307375 307376 307377 307378 307379
SCTV307307EA 5350 CZK
SCTV307307 SCTV307310 SCTV307328 SCTV307367 SCTV307368 SCTV307369 SCTV307370 SCTV307371 SCTV307372 SCTV307373 SCTV307324 SCTV307355 SCTV307366 SCTV307374 SCTV307375 SCTV307376 SCTV307377 SCTV307378 SCTV307379
HPLC waste-sets, V2.0
Chromatografické potřeby Chromatography Spare Parts
The HPLC waste-sets, V2.0 are economical combinations for clean and safe disposal of liquid laboratory waste by HPLC systems or other analytical instruments. All sets consist of a canister and an original SCAT SafetyCap with exhaust filter as well as fittings, blind plugs and a tube connector.
  • Customised system with high-quality set components
  • Safety caps from the premium manufacturer SCAT
  • Safety canisters made of HDPE or PP material
  • Attractive price compared to single purchase
  • Ready to use, no further components necessary
SCAT offers a completeMore Product Information