Tisk…
HPLC supply-sets, V2.0
Dodavatel: S.C.A.T.
307300 307301 307303 307304 307305 307306 307312 307313 307314 307347
SCTV307300EA 3360 CZK
SCTV307300 SCTV307301 SCTV307303 SCTV307304 SCTV307305 SCTV307306 SCTV307312 SCTV307313 SCTV307314 SCTV307347
HPLC supply-sets, V2.0
Chromatografické potřeby Chromatography Spare Parts
The HPLC Supply Sets, V2.0 of the premium manufacturer SCAT are attractively priced combinations of equipment for contamination-free supply of HPLC systems with solvents. The sets each consist of a laboratory bottle with GL 45 thread, a SCAT SafetyCap as well as matching capillaries, suction filters and air valves.
  • Customised system with high quality set components
  • SafetyCaps of the premium manufacturer SCAT
  • Laboratory bottles of DURAN® brand
  • Price attractive in comparison with single purchase
  • Ready for use, no further components necessary
SCAT offersMore Product Information