Tisk…
Safety caps and safety waste cap accessories and replacement parts
Dodavatel: S.C.A.T.
107991 107992 107993 107994 107995 107996 108021 108022 108029 160130 160132 160134 107093 107094 107095 107096 107097 107099 107621 107622 107624 107027 107021 107022 107023 107024 107025 107026 107801 107811 107812 107813 107821 107822 107825 107826 117030 117094 117095 107090 107091 107092 160137 160138 160503 160139 107014 160141 107016 107080 107082 107086 160515 160142 160143 107627 107079 107078 107074 107084 107028 107017 107015 107072 107089 107085 160129 117091 108058 117818 108060 108061 108066 199010 160146 117808 108081 108074 107885 160128 108145 108146 108147 107135 108072 107056 108279 117819 117821 107131 108325 107892 160526 160528 450130 108118
590-0683EA 2020 CZK
590-0683 590-0684 590-0685 590-0686 590-0687 590-0688 590-0711 590-0712 590-0719 590-0815 590-0817 590-0818 590-0530 590-0531 590-0532 590-0533 590-0534 590-0536 590-0556 590-0557 590-0559 590-0514 590-0508 590-0509 590-0510 590-0511 590-0512 590-0513 590-0579 590-0588 590-0589 590-0590 590-0598 590-0599 590-0602 590-0603 590-0806 590-0807 590-0808 590-0858 590-0859 590-0860 590-0892 590-0893 590-0895 590-0894 554-2681 554-2708 554-2683 554-2691 554-2692 554-2696 SCTV160515 554-2709 554-2710 554-4689 554-2690 554-2689 554-2687 554-2694 554-2686 554-2684 554-2682 SCTV107072 554-2699 554-2695 554-2707 SCTV117091 SCTV108058 SCTV117818 SCTV108060 554-1851 554-1856 SCTV199010 590-0008 554-4729 SCTV108081 SCTV108074 590-0031 SCTV160128 SCTV108145 SCTV108146 SCTV108147 SCTV107135 590-0034 SCTV107056 SCTV108279 SCTV117819 SCTV117821 SCTV107131 SCTV108325 SCTV107892 SCTV160526 SCTV160528 SCTV450130 SCTV108118
Safety caps and safety waste cap accessories and replacement parts
Uzávěry Uzávěry bezpečnostní a uzávěry bezpečnostní na odpad
Helpful supplies for the safe use of SCAT and SymLine products safety caps and safety waste caps.
  • SCAT connectors are available in the following versions:
  • Conical connectors
  • Y-connectors
  • Tube connectors
  • Blind plugs
  • Capillary connector
SCAT thread adapters are ideal for connecting safety caps, safety waste caps, funnels, canisters, or other disposal and supply units.