Tisk…
Round canisters
Dodavatel: S.C.A.T.
107704 107706 107711 107722 107712 107721 107720 107730
590-0572EA 2860 CZK
590-0572 590-0573 SCTV107711 SCTV107722 SCTV107712 SCTV107721 SCTV107720 SCTV107730
Round canisters
Nádoby bezpečnostní
SCAT round canisters are ideal for collecting and disposing of smaller quantities of solvents due to their compact dimensions. Several SCAT round canisters can be easily stationed at the HPLC system and equipped with SCAT SafeyWasteCaps.
  • Made of either robust HDPE or PP and therefore resistant to all solvents commonly used in the laboratory
  • Available in the common threads S65, S70/71 and B83
  • Filling capacities range from very compact 2 to 4 L, 10, 25, 50 and 60 L for the collection of voluminous solvents
  • Many individual features such as carrying handles,
More Product Information