Tisk…
Tubes and connectors, SymLine®
Dodavatel: S.C.A.T.
106430 106440 106471 106437 106421 106426 106436 106435 106475 106476 106427 106456 106609 107125 106457 106458 106605 106608 106611 106449
554-4366EA 3030 CZK
554-4366 554-4370 SCTV106471 554-4369 554-4362 554-4365 554-4368 554-4367 SCTV106475 SCTV106476 SCTV106427 SCTV106456 SCTV106609 SCTV107125 SCTV106457 SCTV106458 SCTV106605 SCTV106608 SCTV106611 SCTV106449
Tubes and connectors, SymLine®
Fitinky a konektory
Place the collecting container/canister where needed; the pipe system can be exactly configured to suit specific needs.
  • Straight pipes are available in standard lengths of 300, 900 and 1200 mm
  • Angled pipes are available in the further lengths of 150 and 500 mm
  • A wide range of connectors, branches, and manifolds allow the pipe system to be connected precisely to the conditions in the laboratory
The SymLine® pipe system provides the connection to the central collection point.