Tisk…
Connectors, tubing, stainless steel, 4-port, GL 45, DURAN®
Dodavatel: DWK Life Sciences
292612907
SCOT292612907EA 11220 CZK
SCOT292612907
Connectors, tubing, stainless steel, 4-port, GL 45, DURAN®
Fitinky a konektory
The four-port connector cap is manufactured using three-dimensional metal printing. This method allows the complex form to be optimised using computer aided design software, as well as significantly reducing the amount of metal required for construction. The transfer connection cap can be used with all sizes of DURAN® GL 45 bottles, and facilitates the movement of liquids within a closed and aseptic system.
  • Ideal for chemistry or life science laboratories
  • Sterile liquid transfer between containers
  • Liquid sampling into bottles from larger containers