Tisk…
Adapter, stopper type, DURAN®
Dodavatel: DWK Life Sciences
286576503 286576803 286576109 286576306 286576709 286576906
SCOT286576503EA 11520 CZK
SCOT286576503 SCOT286576803 SCOT286576109 SCOT286576306 SCOT286576709 SCOT286576906
Adapter, stopper type, DURAN®
Adaptéry Adaptéry s kónickým zábrusů
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Adapters, stopper type, solid, socket and cone.