Tisk…
Standard cones with tip, finished, DURAN®
Dodavatel: DWK Life Sciences
286163007 286163401 286164003 286165205 286166004 286168605 286169404 286169601 286170208 286170808 286171007 286171204 286171607 286164406 286172209 286166801
SCOT286163007EA 2240 CZK
SCOT286163007 SCOT286163401 SCOT286164003 SCOT286165205 SCOT286166004 SCOT286168605 SCOT286169404 SCOT286169601 SCOT286170208 SCOT286170808 SCOT286171007 SCOT286171204 SCOT286171607 SCOT286164406 SCOT286172209 SCOT286166801
Standard cones with tip, finished, DURAN®
Adaptéry Adaptéry s kónickým zábrusů
DURAN®, borosilicate glass 3,3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes