Tisk…
DURAN® Burettes, Straight PTFE Stopcock, Class AS, Amber
Dodavatel: DWK Life Sciences
243362708 243363301 243363601 243363901
SCOT243362708EA 7480 CZK
SCOT243362708 SCOT243363301 SCOT243363601 SCOT243363901
DURAN® Burettes, Straight PTFE Stopcock, Class AS, Amber
Byrety
Burettes with PTFE key and straight standard ground joint, made of highly chemically resistant borosilicate glass 3.3. Their precise scaling makes them particularly suitable for titrimetric purposes.
  • UV protection through amber glass
  • The volume content tolerances conform to accuracy class AS
  • Easy-to-read scale with graduation marks to estimate the content
  • Waiting time: 30 seconds
  • Fired-on, highly durable white ceramic
The quantity of liquid required for titration, which is unknown beforehand, can be precisely read off once the reaction is complete.