Tisk…
DURAN® Burettes with Straight Standard Ground Joint, Amber
Dodavatel: DWK Life Sciences
243262701 243263303 243263603 243263903
SCOT243262701EA 6220 CZK
SCOT243262701 SCOT243263303 SCOT243263603 SCOT243263903
DURAN® Burettes with Straight Standard Ground Joint, Amber
Byrety
The original DURAN® burettes are made of highly chemically resistant borosilicate glass 3.3. Their precise scaling makes them particularly suitable for titrimetric purposes.
  • UV protection through amber glass
  • Precise scale ideal for titration
  • Waiting time: 30 seconds
  • With circular ring graduations, straight standard ground stopcock
  • Fired-on, highly durable white print
The amount of liquid required for titration, but previously unknown, can be read off very accurately after the reaction has been completed. Adjustment is made to drain ('Ex') for 20 °CMore Product Information