Tisk…
Láhve, úzkohrdlé, se standardní broušenou zátkou, DURAN®
Dodavatel: DWK Life Sciences
211647301 211648606 211649105 211643608 211642406 211644407 211645403 211646305 211641701
SCOT211647301EA 6590 CZK
SCOT211647301 SCOT211648606 SCOT211649105 SCOT211643608 SCOT211642406 SCOT211644407 SCOT211645403 SCOT211646305 SCOT211641701
Láhve, úzkohrdlé, se standardní broušenou zátkou, DURAN®
Láhve Láhve skleněné
DURAN®, borosilicate glass 3.3, clear or brown, with standard ground joint, without stopper.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Glass Type I according to USP, EP and JP
Brown tinted to protect light-sensitive substances.
With printed retrace code (batch identification).