Tisk…
Glass tubings
Dodavatel: Schott AG
1193963 1193962 1193964 1193985 1193986 1193988 1194016 1194017 1194018 1194020 1391244 1402201 1404444 1028081 1485595 1487826 1504772 1270387 1518580 1518606 1028943 1027897 1028162 1564520 1263533 1545315 1545822 1579276 1391200 1552454 1552451
SCOR1193963EA 7860 CZK
SCOR1193963 SCOR1193962 SCOR1193964 SCOR1193985 SCOR1193986 SCOR1193988 SCOR1194016 SCOR1194017 SCOR1194018 SCOR1194020 SCOR1391244 SCOR1402201 SCOR1404444 SCOR1028081 SCOR1485595 SCOR1487826 SCOR1504772 SCOR1270387 SCOR1518580 SCOR1518606 SCOR1028943 SCOR1027897 SCOR1028162 SCOR1564520 SCOR1263533 SCOR1545315 SCOR1545822 SCOR1579276 SCOR1391200 SCOR1552454 SCOR1552451
Glass tubings
Hadičky Přenosová hadička
AR-Glas®, clear.
  • Good chemical and physical properties
  • Suitable for the production of items that must normally withstand only brief exposure to aggressive chemicals
  • Not resistant to high thermal loads
Tube length ~1500 mm.