Tisk…
DURAN® Stirring Rods, Glass
Dodavatel: Schott AG
1026637 1026631 1026554 1026556 1026557 1026558 1026559 1026560 1026562 1026563 1026564 1026566 1026633 1026641 1026674 1027002 1027003 1027004
SCOR1026637EA 28900 CZK
SCOR1026637 SCOR1026631 SCOR1026554 SCOR1026556 SCOR1026557 SCOR1026558 SCOR1026559 SCOR1026560 SCOR1026562 SCOR1026563 SCOR1026564 SCOR1026566 SCOR1026633 SCOR1026641 SCOR1026674 SCOR1027002 SCOR1027003 SCOR1027004
DURAN® Stirring Rods, Glass
Míchadla Tyčky míchací
Very long glass rods made from Borosilicate 3.3 glass.
  • High chemical resistance
  • High hydrolytical resistance
  • High temperature resistance
  • High resistance to thermal shock