Tisk…
Headspace oven and sample changer for coulometric KF titration
Dodavatel: SI Analytics
285227850 285227820 285227840
SCOI285227850EA 674400 CZK
SCOI285227850 SCOI285227820 SCOI285227840
Headspace oven and sample changer for coulometric KF titration
Titrátory
The headspace oven technique of the TO 7280 allows the separation of the water to be titrated from liquid, solid and semi-solid (paste-like) samples. In combination with the TW 7650 sample changer, up to 49 samples can be analysed.
  • Semi-automatic or fully automatic water determination
  • No contamination of the anolyte, generator and indicator electrodes
  • Application range from 35 to 280 °C
  • Reproducibility: ±3 μg bei 10 to 1000 μg, 0,3% bei >1 mg
Requires TitriSoft version 3.50 or higher to operate. One position is reserved for the zero-vial toMore Product Information