Tisk…
pH combination electrodes, ProcessLine
Dodavatel: SI Analytics
285113700 285113490 285113550 285113560 285113570 285113580 285113590 285113650 285113660 285113670 285113680 285113690 285113710 285113720 285113740 285113750 285113770 285113780 285113790 285113800 285113505 285113515 285113525 285113535 285113565 285113575 285113605 285114505 285114570 285114605 285114610 285114555 285114480 285114525 285114530 285114535 285114575 285114580 285114630 285114660 285114615 285114650 285114655 285114510 285114520 285114560 285114625 285114635 285114620
662-1702EA 12780 CZK
662-1702 662-1691 662-1692 662-1693 662-1694 662-1695 662-1696 662-1697 662-1698 662-1699 662-1700 662-1701 662-1703 662-1704 662-1705 662-1706 662-1708 662-1709 662-1710 662-1711 SCOI285113505 SCOI285113515 SCOI285113525 SCOI285113535 SCOI285113565 SCOI285113575 SCOI285113605 SCOI285114505 SCOI285114570 SCOI285114605 SCOI285114610 SCOI285114555 SCOI285114480 SCOI285114525 SCOI285114530 SCOI285114535 SCOI285114575 SCOI285114580 SCOI285114630 SCOI285114660 SCOI285114615 SCOI285114650 SCOI285114655 SCOI285114510 SCOI285114520 SCOI285114560 SCOI285114625 SCOI285114635 SCOI285114620
pH combination electrodes, ProcessLine
Elektrody Elektrody pH
These electrodes are low maintenance sensors for heavy duty process applications in the chemical industry. They are ideally suitable for measuring media with extreme ionic strength - whether boiler feed water or brine - also in strongly oxidizing acid and alkali containing media.
  • Low-maintenance electrodes: no electrolyte topping up or installation of complicated pressure sequence control required
  • Pinhole diaphragms, so reference electrode is not polluted or blocked
  • Duralid electrolyte with high KCl content combines very long working life with rapid and
More Product Information