Tisk…
Serological pipettes, Shorty, KIMBLE®
Dodavatel: DWK Life Sciences
SP72115-50510 SP72115-5110
SCERSP72115-50510EA 32990 CZK
SCERSP72115-50510 SCERSP72115-5110
Serological pipettes, Shorty, KIMBLE®
Pipety Pipety serologické
Shorty pipettes are made from glass and are ideal for use in tissue culture work and other procedures performed under a fume hood or in other confined areas.
  • 'Reverse' and 'negative' graduations increase the versatility of the pipette
  • Calibrated 'to deliver' with blow-out after free flow of liquid has ended
  • Multi-pack bag: One or several pipettes can be removed without contaminating the entire pack
  • Non pyrogenic, sterile, disposable
Balení: Individually wrapped in paper/plastic or packed in a multi-pack bag.