Tisk…
Filter holder, stainless steel
Dodavatel: Sartorius
16213
SART16213EA 17790 CZK
SART16213
Filter holder, stainless steel
Zařízení filtrační
Stainless steel closed filter holder for microbiological, analytical sampling and testing of gas in pressurised lines.
  • Streamlined design with ideal flow path geometry
  • Can be sterilised in a hot air oven
Informace o dodávce: The filter holder consists of a funnel shaped base with a hose nipple, a metal sintered frit as a filter support and a sealing ring, as well as a funnel shaped top part with a hose nipple and a back pressure screen.