Tisk…
Ultrafiltration membrane filters, PES
Dodavatel: Sartorius
14629-47-D 14639-47-D 14650-47-D 14659-47-D 14679-47-D 14629-25-D 14639-25-D 14659-25-D 14650-25-D 14609-76-D 14629-76-D 14639-76-D 14650-76-D 14679-76-D 14609-47-D 14609-63-D 14629-63-D 14639-63-D 14659-63-D 14668-63-D 14668-47-D
SART14629-47-DEA 4710 CZK
SART14629-47-D SART14639-47-D SART14650-47-D SART14659-47-D SART14679-47-D SART14629-25-D SART14639-25-D SART14659-25-D SART14650-25-D SART14609-76-D SART14629-76-D SART14639-76-D SART14650-76-D SART14679-76-D SART14609-47-D SART14609-63-D SART14629-63-D SART14639-63-D SART14659-63-D SART14668-63-D SART14668-47-D
Ultrafiltration membrane filters, PES
Filtry Filtry membránové
These are general purpose membrane that provide excellent performance with most solutions when retentate recovery is of primary importance.
  • No hydrophobic or hydrophilic interactions
  • Low fouling characteristics, exceptional flux and broad pH range
Objednat
Produktová specifikace
Environmentally Preferable
Tloušťka 120 μm