Tisk…
Membrane filters with dehydrated media pads for microbiological control
Dodavatel: Sartorius
14051-47-N 14056-47-N 14058-47-N 14059-47-N 14063-47-N 14064-47-N 14066-47-N 14068-47-N 14069-47-N 14070-47-N 14074-47-N 14079-47-N 14084-47-N 14051-50-N 14053-47-N 14053-50-N 14055-47-N 14055-50-N 14056-50-N 14058-50-N 14059-50-N 14062-47-N 14062-50-N 14063-50-N 14064-50-N 14066-50-N 14068-50-N 14069-50-N 14070-50-N 14072-50-N 14074-50-N 14075-47-N 14075-50-N 14078-50-N 14079-50-N 14097-50-N 14084-50-N 14087-47-N 14087-50-N 14078-47-N 14086-47-N 14080-47-N 14080-50-N 14090-47-N 14096-50-N 14091-47-N 14090-50-N 14076-47-N 14083-50-N 14089-47-N 14097-47-N 14091-50-N 14076-50-N 14096-47-N 14077-47-N 14049-47-N
391-3904EA 6790 CZK
391-3904 391-3905 710-1103 710-1104 391-0235 710-1106 710-1108 710-1110 391-0237 710-1111 391-0240 710-1114 391-0236 516-7000 391-3900 516-7001 391-3902 391-3955 391-3956 391-3958 391-3959 391-3962 516-7006 516-7007 516-7008 391-3966 516-7009 391-3969 391-3970 516-7012 516-7013 391-3901 391-3975 391-3978 391-3979 391-3997 516-0217 710-1117 710-1100 710-1113 710-1116 710-1115 SART14080-50-N 391-3907 710-1102 710-1101 SART14090-50-N 710-1118 SART14083-50-N 710-1121 710-0844 SART14091-50-N SART14076-50-N SART14096-47-N SART14077-47-N SART14049-47-N
Membrane filters with dehydrated media pads for microbiological control
Média mikrobiologická Připravená mikrobiologická média
Each set consists of a Ø 50 mm or Ø 47 mm pad, impregnated with nutrient media, packed dry and sterile in a Petri dish, with a membrane filter. After filtration of the sample the membrane is placed in the dish and incubated. More types available on request.
  • Simply prepared by the addition of 3,5 ml sterile, demineralised water
  • Individually sterile packed
  • Can be kept 18 to 24 months at room temperature, depending on the type