Tisk…
Membrane filters, type 113
Dodavatel: Sartorius
11301-100-N 11301-37-N 11304-100-G 11304-37-N 11305-100-N 11306-100-N 11306-85-N 11306-90-N 11301-100-G 11301-25-N 11301-47-ACN 11301-47-N 11301-50-N 11302-13-N 11302-25-N 11302-47-ACN 11302-47-N 11302-50-N 11303-100-G 11303-25-N 11303-47-ACN 11303-47-N 11303-50-N 11303-90-G 11304-13-N 11304-25-N 11304-47-ACN 11304-47-N 11304-50-ACN 11304-50-N 11304-90-G 11305-142-G 11305-25-N 11305-47-N 11305-50-N 11306-100-G 11306-13-N 11306-25-N 11306-37-N 11306-47-ACN 11306-47-N 11306-50-ACN 11306-50-N 11306-90-G 11342-13-N 11342-25-N 11342-47-N 11342-50-N 11301-70-G 11301-13-N 11301-50-ACN 11305-50-ACN 11342-90-G 11302-90-G 11305-13-N 11305-47-ACN 11327-25-N 11327-47-N
SART11301-100-NEA 19700 CZK
SART11301-100-N 514-0752 SART11304-100-G 512-5842 SART11305-100-N SART11306-100-N 611-0677 514-0754 512-5510 512-5210 512-5861 512-5310 512-5410 512-5112 512-5212 512-5862 512-5312 512-5412 512-5513 512-5213 512-5863 512-5313 512-5413 512-5914 512-5114 512-5214 512-5864 512-5314 512-5874 512-5414 512-5915 512-5615 512-5215 512-5315 512-5415 512-5516 512-5116 512-5216 512-5843 611-0675 512-5316 512-5876 512-5416 512-5917 512-5111 512-5211 512-5311 512-5411 SART11301-70-G 512-5110 512-5871 512-5875 SART11342-90-G SART11302-90-G SART11305-13-N SART11305-47-ACN SART11327-25-N SART11327-47-N
Membrane filters, type 113
Filtry Filtry membránové
Cellulose nitrate hydrophilic membrane filters. For sample pretreatment, particle testing and chemotaxis.
* Sterile