Tisk…
Calibration weights, OIML class F1, Proof Line
Dodavatel: Sartorius Balances
YCW413-00 YCW513-02 YCW413-02 YCW023-02 YCW153-00 YCW153-02 YCW213-00 YCW213-02 YCW023-00 YCW013-00 YCW013-02 YCW223-00 YCW423-00 YCW553-00 YCW353-00 YCW313-00 YCW323-02 YCW723-02 YCW653-00 YCW653-02 YCW713-02 YCW223-02 YCW123-00 YCW053-00 YCW113-02 YCW123-02 YCW313-02 YCW353-02 YCW513-00 YCW523-00 YCW723-00 YCW113-00 YCW253-00 YCW053-02 YCW253-02 YCW613-02 YCW423-02 YCW523-02 YCW623-02 YCW453-02 YCW553-02 YCW613-00 YCW323-00 YCW623-00 YCW453-00 YCW713-00 YCW313-AC-02 YCW353-AC-02 YCW353-AC-00 YCW313-AC-00 YCW323-AC-02 YCW323-AC-00 YCW623-AC-02 YCW623-AC-00 YCW653-AC-00 YCW613-AC-02 YCW613-AC-00 YCW713-AC-02 YCW713-AC-00 YCW653-AC-02 YCW553-AC-00 YCW423-AC-00 YCW453-AC-02 YCW453-AC-00 YCW423-AC-02 YCW413-AC-02 YCW413-AC-00 YCW523-AC-02 YCW553-AC-02 YCW513-AC-02 YCW513-AC-00 YCW523-AC-00
SARBYCW413-00EA 2870 CZK
SARBYCW413-00 SARBYCW513-02 SARBYCW413-02 SARBYCW023-02 SARBYCW153-00 SARBYCW153-02 SARBYCW213-00 SARBYCW213-02 SARBYCW023-00 SARBYCW013-00 SARBYCW013-02 SARBYCW223-00 SARBYCW423-00 SARBYCW553-00 SARBYCW353-00 SARBYCW313-00 SARBYCW323-02 SARBYCW723-02 SARBYCW653-00 SARBYCW653-02 SARBYCW713-02 SARBYCW223-02 SARBYCW123-00 SARBYCW053-00 SARBYCW113-02 SARBYCW123-02 SARBYCW313-02 SARBYCW353-02 SARBYCW513-00 SARBYCW523-00 SARBYCW723-00 SARBYCW113-00 SARBYCW253-00 SARBYCW053-02 SARBYCW253-02 SARBYCW613-02 SARBYCW423-02 SARBYCW523-02 SARBYCW623-02 SARBYCW453-02 SARBYCW553-02 611-1707 SARBYCW323-00 611-1708 SARBYCW453-00 SARBYCW713-00 SARBYCW313-AC-02 SARBYCW353-AC-02 SARBYCW353-AC-00 SARBYCW313-AC-00 SARBYCW323-AC-02 SARBYCW323-AC-00 SARBYCW623-AC-02 SARBYCW623-AC-00 SARBYCW653-AC-00 SARBYCW613-AC-02 SARBYCW613-AC-00 SARBYCW713-AC-02 SARBYCW713-AC-00 SARBYCW653-AC-02 SARBYCW553-AC-00 SARBYCW423-AC-00 SARBYCW453-AC-02 SARBYCW453-AC-00 SARBYCW423-AC-02 SARBYCW413-AC-02 SARBYCW413-AC-00 SARBYCW523-AC-02 SARBYCW553-AC-02 SARBYCW513-AC-02 SARBYCW513-AC-00 SARBYCW523-AC-00
Calibration weights, OIML class F1, Proof Line
Závaží Závaží kalibrační
Proof Line weights are high quality working standards for everyday use. These weights are made of high quality material with polished or precision lathed surfaces.
  • 0 to 5 mg leaf weights: Aluminium
  • 10 to 500 mg leaf weights: Nickel silver
  • 1 g to 10 Kg knob weights: Stainless steel
Certifikace: Compliance with OIML R111-1.