Tisk…
Precision (milligram) laboratory balances, Cubis® II MCE
Dodavatel: Sartorius Balances
MCE10202S2S000 MCE1203S2S00-A MCE1202S2S00-0 MCE623S2S00-R MCE5203S2S00-E MCE5203P2S00-E MCE3203S2S00-E MCE2203S2S00-E MCE2203P2S00-E MCE1203S2S00-E MCE623S2S00-E MCE623P2S00-E MCE323S2S00-E MCE5203S2S00-R MCE5203P2S00-R MCE3203S2S00-R MCE2203S2S00-R MCE2203P2S00-R MCE1203S2S00-R MCE623P2S00-R MCE323S2S00-R MCE5202S2S00-E MCE5202S2S00-R MCE14202S2S000 MCE14202P2S000 MCE12201S2S000 MCE11201S2S000 MCE5203S2S00-A MCE5203P2S00-A MCE3203S2S00-A MCE2203S2S00-A MCE2203P2S00-A MCE623S2S00-A MCE623P2S00-A MCE323S2S00-A MCE5203S2S00-I MCE5203P2S00-I MCE3203S2S00-I MCE2203S2S00-I MCE2203P2S00-I MCE1203S2S00-I MCE623S2S00-I MCE623P2S00-I MCE323S2S00-I MCE5203S2S00-U MCE5203P2S00-U MCE3203S2S00-U MCE2203S2S00-U MCE2203P2S00-U MCE1203S2S00-U MCE623S2S00-U MCE623P2S00-U MCE323S2S00-U MCE6202S2S00-0 MCE6202P2S00-0 MCE4202S2S00-0 MCE8201S2S00-0 MCE5202S2S00-A MCE5202S2S00-I MCE5202S2S00-U MCE14202S-2CEU MCE14202S-2S00 MCE12201S2CFR0
SARBMCE10202S2S000EA 262720 CZK
SARBMCE10202S2S000 SARBMCE1203S2S00-A SARBMCE1202S2S00-0 SARBMCE623S2S00-R SARBMCE5203S2S00-E SARBMCE5203P2S00-E SARBMCE3203S2S00-E SARBMCE2203S2S00-E SARBMCE2203P2S00-E SARBMCE1203S2S00-E SARBMCE623S2S00-E SARBMCE623P2S00-E SARBMCE323S2S00-E SARBMCE5203S2S00-R SARBMCE5203P2S00-R SARBMCE3203S2S00-R SARBMCE2203S2S00-R SARBMCE2203P2S00-R SARBMCE1203S2S00-R SARBMCE623P2S00-R SARBMCE323S2S00-R SARBMCE5202S2S00-E SARBMCE5202S2S00-R SARBMCE14202S2S000 SARBMCE14202P2S000 SARBMCE12201S2S000 SARBMCE11201S2S000 SARBMCE5203S2S00-A SARBMCE5203P2S00-A SARBMCE3203S2S00-A SARBMCE2203S2S00-A SARBMCE2203P2S00-A SARBMCE623S2S00-A SARBMCE623P2S00-A SARBMCE323S2S00-A SARBMCE5203S2S00-I SARBMCE5203P2S00-I SARBMCE3203S2S00-I SARBMCE2203S2S00-I SARBMCE2203P2S00-I SARBMCE1203S2S00-I SARBMCE623S2S00-I SARBMCE623P2S00-I SARBMCE323S2S00-I SARBMCE5203S2S00-U SARBMCE5203P2S00-U SARBMCE3203S2S00-U SARBMCE2203S2S00-U SARBMCE2203P2S00-U SARBMCE1203S2S00-U SARBMCE623S2S00-U SARBMCE623P2S00-U SARBMCE323S2S00-U SARBMCE6202S2S00-0 SARBMCE6202P2S00-0 SARBMCE4202S2S00-0 SARBMCE8201S2S00-0 SARBMCE5202S2S00-A SARBMCE5202S2S00-I SARBMCE5202S2S00-U 611-5267 611-5268 SARBMCE12201S2CFR0
Precision (milligram) laboratory balances, Cubis® II MCE
Váhy
Cubis® II MCE precision balances with essential user interface offer a maximum load between 320 and 14200 g and a readability of 1 to 100 mg provide, ideal model for every application.
  • Modular premium balances with thousands of hardware and software configurations and adaptable software
  • Seamless connectivity
  • Long-term flexibility
  • Submission-ready data
  • Governance through user management
  • Integration of SOPs
  • Intuitive applications
  • Status centre notifications
Weighing in a regulated environment can be complex, with stringent compliance directives andMore Product Information