Tisk…
Drums, Stainless Steel, with Toggle Clamps
Dodavatel: Sampling Systems
A626-1 A626-2 A626-3 A626-5 A626-6 A626-8 A626-10 A626-12 A626-15
SAMPA626-1EA 38300 CZK
SAMPA626-1 SAMPA626-2 SAMPA626-3 SAMPA626-5 SAMPA626-6 SAMPA626-8 SAMPA626-10 SAMPA626-12 SAMPA626-15
Drums, Stainless Steel, with Toggle Clamps
Sudy, soudky, bubny
This popular container range is available in a range of sizes from 1 to 15 L. They are manufactured from 316L stainless steel and finished to a high quality finish. Ideal for work in a range of different environments include small scale production, laboratories and general day to day use.
  • They are ideal for working with both powders and liquids
  • The containers come with a drop-on lid
  • The lid is secured with toggle clamps and an O-ring seal
  • Simply pull down the toggle clamps to secure the lid
  • Storage conditions: Dry and ambient temperature