Tisk…
Pointed Scissors
Dodavatel: Sampling Systems
A364-300 A364-150
SAMPA364-300EA 1410 CZK
SAMPA364-300 SAMPA364-150
Pointed Scissors
Nůžky
Surgical grade scissors for cutting.
  • Made from surgical grade 420 stainless steel
  • Great for cutting and manipulating samples
These high quality pointed scissors are made from surgical grade 420 stainless steel and great for cutting and manipulating.