Tisk…
Double Ended Spoons
Dodavatel: Sampling Systems
A104-210 A104-250 A104-300 A104-150 A104-180
SAMPA104-210EA 788 CZK
SAMPA104-210 SAMPA104-250 SAMPA104-300 SAMPA104-150 SAMPA104-180
Double Ended Spoons
Lžíce
Double ended spoon for taking different sample sizes.
  • Made from stainless steel
  • Ideal for sampling powders and liquids
  • Various sizes available
These double ended spoons are great for taking different sample sizes using the same spoon. The lengths and volume capacities of the spoons vary allowing for flexibility in sampling both powders and liquids.