Tisk…
Powder funnels, USP Class VI, double-bagged, ULTRA
Dodavatel: Sampling Systems
9111H-200S
SAMP9111H-200SEA 6100 CZK
SAMP9111H-200S
Powder funnels, USP Class VI, double-bagged, ULTRA
Nálevky Násypky
The SteriWare Ultra range is the premium range from Sampling Systems. These powder funnels are double bagged and are made from high quality USP class VI HDPE. A box liner is also used giving a third layer of protection.
  • Double-bagged
  • Produced in class 100,000 cleanroom, packed in Class 10,000 cleanroom
  • Crevice free with smooth finish
  • Angle of the cone is designed to give optimum product flow
These sterile funnels are designed to be used straight from the bag. The double bagging means that the outer bag can be removed whilst entering the cleanroom,More Product Information