Tisk…
Scoops, USP Class VI, double-bagged, ULTRA
Dodavatel: Sampling Systems
9045H-1000S 9045H-125S 9045H-250S
SAMP9045H-1000SEA 8120 CZK
SAMP9045H-1000S SAMP9045H-125S SAMP9045H-250S
Scoops, USP Class VI, double-bagged, ULTRA
Lopatky
The double bagged sterile scoops are made from high quality USP class VI HDPE. A box liner is also used giving a third layer of protection.
  • Double-bagged
  • Produced in Class 100,000 cleanroom, packed in Class 10,000 cleanroom
  • Sterilised by gamma irradiation
These scoops all have an ergonomic handle which is positioned high up the body so as to keep the hand as far away from the powder as possible.The scoops are designed to be used straight from the bag. The double bagging means that the outer bag can be removed whilst entering the cleanroom, ensuringMore Product Information