Tisk…
Re-Usable Cleaning Coupons
Dodavatel: Sampling Systems
8644SIL-100 8644H-100 8644PP-100 8644AB-100 8644N-100 8644S6-50S 8644S4-100S 8644S4-50B 8644S4-50S 8644AL-100 8644AB-50 8644SIL-50 8644PVC-100 8644E-100
SAMP8644SIL-100EA 7170 CZK
SAMP8644SIL-100 SAMP8644H-100 SAMP8644PP-100 SAMP8644AB-100 SAMP8644N-100 SAMP8644S6-50S 300-0609 300-0610 300-0611 SAMP8644AL-100 SAMP8644AB-50 SAMP8644SIL-50 SAMP8644PVC-100 SAMP8644E-100
Re-Usable Cleaning Coupons
Ubousky/Utěrky Podložky čistící
Cleaning coupons are great for validating a cleaning regime.
  • All popular sizes available
  • Wide variety of materials and finishes
Sampling Systems' extensive range of cleaning coupons come in a diverse selection of materials, sizes, and finishes. They are ideal for validating cleaning regimes. Popular materials include 316 stainless steel, PTFE, aluminium, and HDPE.