Tisk…
VWR®, Pre-weighed dehydrated culture media in sachets
Dodavatel: VWR Chemicals
S246SAC5EA 569 CZK
S246SAC5 A887SAC5 S378SAC5 B202SAC5 B455SAC5 A990SAC5 A935SAC5 B453SAC5 A484SAC5 A265SAC5 A919SAC5 A857SAC5 B857SAC5 A865SAC5 B923SAC5 A704SAC5 A979SAC5 S821SAC5 B710SAC5 A389SAC5 A971SAC5 A882SAC5 A355SAC5 A925SAC5 A188SAC5 A890SAC5 A869SAC5 B700SAC5 A807SAC5 A836SAC5 A975SAC5 B475SAC5 A128SAC5 A875SAC5 B989SAC5 B982SAC5 A986SAC5 A760SAC5 B206SAC5 B254SAC5 A487SAC5 B872SAC5 A333SAC5 A751SAC5 A961SAC5 B419SAC5 A794SAC5 B901SAC5 A438SAC5 A902SAC5 A700SAC5 A852SAC5 A993SAC5 A940SAC5 A391SAC5 B397SAC5 A167SAC5 B167SAC5 B418SAC5 A415SAC5 B415SAC5 A851SAC5 A427SAC5 A731SAC5 B811SAC5 B833SAC5 B336SAC5 A540SAC5 A723SAC5 A338SAC5 B739SAC5 A336SAC5 A870SAC5 B878SAC5 A317SAC5 B317SAC5 A318SAC5 B318SAC5 B117SAC5 B734SAC5 A315SAC5 S834SAC5 A322SAC5 A817SAC5 A329SAC5 B322SAC5 A253SAC5 A259SAC5 A261SAC5 B261SAC5 S158SAC5 A729SAC5 B150SAC5 A383SAC5 A234SAC5 S460SAC5 A382SAC5 A407SAC5 A193SAC5 A388SAC5 B387SAC5 A387SAC5 A457SAC5 A777SAC5 A912SAC5 A174SAC5 A797SAC5 B706SAC5 B894SAC5 B884SAC5 S358SAC5 A750SAC5 S272SAC5 A830SAC5 A785SAC5 B368SAC5 S365SAC5 A304SAC5 A709SAC5 A748SAC5 A499SAC5 B499SAC5 B396SAC5 A483SAC5 B494SAC5 A491SAC5 A190SAC5 A498SAC5 B495SAC5 A433SAC5 A774SAC5
VWR®, Pre-weighed dehydrated culture media in sachets
Média mikrobiologická Mikrobiologická média dehydratovaná
Dehydrated culture media in small sachets ready to make up exactly 500 ml of medium.
  • Quick: Simply open the sachet and you have the right quantity of powder to prepare 500 ml of medium
  • Safe: No more need to transfer and weigh powder
  • Always fresh media for best quality: All the dehydrated media inside the sachet is always used immediately after opening the packaging
  • Space saving: You can have tens of dehydrated media stored in a very small space inside your laboratory
  • Saving money: You buy only the quantity of dehydrated media that you really need
Every pack contains 5 sachets each of which allows to prepare 500 ml of medium.
Informace pro objednávání: About 140 different culture media available.