Tisk…
Clamps, four pronged, universal
Dodavatel: RSG
400.000
RSGA400.000EA 1030 CZK
RSGA400.000
Clamps, four pronged, universal
Svorky Svorky pro běžné účely
Universal clamp made from zinc die-cast.
  • Four-finger jaws
  • Jaws fitted with cork