Tisk…
Stainless steel, wire gauzes with frame pro hořáky plynové
Dodavatel: RSG
247.935 247.955 247.995 247.975
RSGA247.935EA 1640 CZK
RSGA247.935 RSGA247.955 RSGA247.995 RSGA247.975
Stainless steel, wire gauzes with frame pro hořáky plynové
Stainless steel
Wire mesh with frame
  • Stainless steel 1.4301
  • mesh: 0.6 x 2.0 mm
  • fine mesh centre: Heat resistant stainless steel