Tisk…
Microscopy instrument set
Dodavatel: RSG
066.805 066.807
RSGA066.805EA 3590 CZK
RSGA066.805 RSGA066.807
Microscopy instrument set
Sady řezací
Microscopy instrument set in a grey wooden box, all articles are completely made of stainless steel.
  • 1 pair of microscopy scissors
  • 1 microscopy needle straight
  • 1 microscopy needle bent
  • 1 pair of tweezers, pointed straight with guide pin
  • 1 scalpel
Informace pro objednávání: On demand the arrangement of the 7 pcs. set can be individualised.