Tisk…
Crucible tongs, double curved, polished/unpolished tips
Dodavatel: RSG
840.200 840.250 840.300 840.220 040.300 040.250 040.220 040.200
231-1131EA 445 CZK
231-1131 231-1132 231-1133 231-4251 RSGA040.300 RSGA040.250 RSGA040.220 RSGA040.200
Crucible tongs, double curved, polished/unpolished tips
Kleště
These cruscible tongs with double curved unpolished tips are made of stainless steel (Remanit 4301).