Tisk…
Gas monitor for supplied breathing air, GX4
Dodavatel: RPB SAFETY
08-400-G 08-420-01 08-420-02 08-420-03 08-420-04 08-231 08-435 08-436 08-434
RPBS08-400-GEA 0 CZK
RPBS08-400-G RPBS08-420-01 RPBS08-420-02 RPBS08-420-03 RPBS08-420-04 RPBS08-231 RPBS08-435 RPBS08-436 RPBS08-434
Gas monitor for supplied breathing air, GX4
Polomasky a filtrační respirátory Systémy polomasek
The RPB GX4 gas monitor continually monitors the quality of supplied breathing air. As soon as the air is no longer of EN breathing air standard, the unit will alarm to warn the respirator wearer that air entering the respirator is no longer safe to breath.
  • Illuminated display that shows real time data
  • Complete quick and easy calibration checks with no set points required
  • 103 dBA staccato alarm
  • Monitor up to four gases simultaneously
  • Sturdy mounting bracket
  • Cartridges can be easily removed and replaced
  • Robust power inlet
  • Push-release air inlet
  • Strong
More Product Information