Tisk…
Respirators, industrial, PX5 PAPR Series
Dodavatel: RPB SAFETY
03-801-FR-EUUK 03-891 04-854
RPBS03-801-FR-EUUKEA 0 CZK
RPBS03-801-FR-EUUK RPBS03-891 RPBS04-854
Respirators, industrial, PX5 PAPR Series
Polomasky a filtrační respirátory Systémy polomasek
The RPB PX5 is a battery powered air purifying respirator fan unit which draws in contaminated air from the wearer's immediate vicinity, passing it through internal filters, and supplying the filtered air to an approved RPB loose fitting respirator head top.
  • Backpack option available for applications that require a greater range of movement and work across varied angles
  • With a completely sealed housing, the PX5 has been certified to IP 65 rating in use and certified to IP 67 rating with cleaning kit in place
  • Filter door cannot mount to the unit unless a