Tisk…

Tris-(hydroxymethyl)aminomethan hydrochlorid

Dodavatel: Roth Carl

Synonyms: Tris(hydroxymethyl)methylamine hydrochloride, 2-Amino-2-(hydroxymethyl)propane-1,3-diol hydrochloride, Trizma® hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, TRIS HCl, Tromethane hydrochloride, Tris-HCl, Tris(hydroxymethyl)methylammonium chloride, Trometamol hydrochloride, THAM HCl

9090.2
ROTH9090.2EA 0 CZK
ROTH9090.2
Tris-(hydroxymethyl)aminomethan hydrochlorid
Tris-(hydroxymethyl)aminomethan hydrochlorid
Vzorec: NH₂C(CH₂OH)₃·HCl
MW: 157.6
Bod tání: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00012590
CAS číslo: 1185-53-1
EINECS: 214-684-5
Merck Index: 12,09902
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cbdc9446fdb9bcfe3f1d992ff20a16726f98113&vid=3d668801395b81fa595401a752279006da8fd3cb&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR