Tisk…

Anti-IgG Goat Polyclonal Antibody (Biotin)

Dodavatel: Rockland Immunochemicals
618-106-012
ROCK618-106-012EA 8980 CZK
ROCK618-106-012
Anti-IgG Goat Polyclonal Antibody (Biotin)
Protilátky
Anti-Ferret IgG (gamma chain) (GOAT) Antibody is suitable for immunoblotting (western or dot blot), ELISA, and immunochemistry requiring extremely low background levels, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG γ
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: γ chain
Host: Goat
Isotype: IgG
Reactivity: Ferret
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR