Tisk…

Anti-IgA Goat Polyclonal Antibody

Dodavatel: Rockland Immunochemicals
618-101-006
ROCK618-101-006EA 8170 CZK
ROCK618-101-006
Anti-IgA Goat Polyclonal Antibody
Protilátky
Type: Secondary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Ferret
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR