Tisk…

Anti-IgG (H+L) Sheep Polyclonal Antibody (AP (Alkaline Phosphatase))

Dodavatel: Rockland Immunochemicals
612-6502
ROCK612-6502EA 9610 CZK
ROCK612-6502
Anti-IgG (H+L) Sheep Polyclonal Antibody (AP (Alkaline Phosphatase))
Protilátky
This product has been assayed against 1.0 µg of Rat IgG in a standard capture ELISA using pNPP p-nitrophenyl phosphate code # NPP-10 as a substrate for 30 minutes at room temperature.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: Heavy and Light chain
Host: Sheep
Isotype: IgG
Reactivity: Rat
Objednat

Produktová specifikace

Symbol antigenu IgG (H+L)
Druh protilátky Sekundární
Klonovalita Polyclonal
Spojení AP (Alkaline Phosphatase)
Reaktivita Rat
Hostitel Sheep
Western blot Yes
ELISA Yes
Imunochemie Yes
Imunoprecipitace Yes
Posun gelu Yes
Průtoková cytometrie Yes
ChIP Yes
Neutralizace Yes
Imunogen Rat IgG whole molecule
Formát Antibody fraction
Formulář Liquid (sterile filtered)
Čištění purified by immunoaffinity chromatography
Koncentrace 0,62 mg/ml by UV absorbance at 280 nm
Křížová adsorbce none
Skladovací pufr 0,05 M Tris Chloride, 0,15M Sodium Chloride, 0,001M Magnesium Chloride, 0,0001M Zinc Chloride, 50% (v/v) Glycerol
Skladovací teplota Cool
Transportní teplota Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR