Tisk…

Anti-AQP2 Rabbit Polyclonal Antibody

Dodavatel: Rockland Immunochemicals
612-401-D08
ROCK612-401-D08EA 27490 CZK
ROCK612-401-D08
Anti-AQP2 Rabbit Polyclonal Antibody
Protilátky
Anti-Aquaporin 2 pS261 antibody is suitable for use in ELISA, Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 29 kDa AQP2 Protein phosphorylated at Ser261 in the appropriate cell lysate.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:1000

Type: Primary
Antigen: AQP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR